Learn More
Medchemexpress LLC 2,2'-bipyridine-d8 | 32190-42-4 | 99.3% | 164.24 g/mol | C10D8N2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYW4404095MG
Encompass_Preferred
2,2′-Bipyridine-d8 is the deuterium-labeled isotopologue of 2,2′-bipyridine supplied for research use. It is intended as an analytical standard and tracer for quantitative analysis by NMR, GC-MS, and LC-MS, and is provided with supporting documentation for laboratory use.
  • High chemical purity (99.3%).
  • Isotope-labeled tracer suitable as an internal standard for NMR, GC-MS, and LC-MS.
  • Available in small milligram quantities for analytical workflows.
  • Includes datasheet, safety data sheet, and certificate of analysis.
  • Intended for research use only; not for human or clinical applications.
Catalog No. 50-002-81361