Learn More
Medchemexpress LLC 2,2'-bipyridine-d8 | 32190-42-4 | MFCD00190432 | 99.3% | 164.24 g·mol⁻¹ | C10D8N2 | 10 MG

Supplier: Medchemexpress LLC HYW44040910MG
2,2′-Bipyridine-d8 is the deuterium-labeled isotopologue of 2,2′-bipyridine supplied as a white to off-white solid for analytical and synthetic chemistry. It is intended for use as a tracer or internal standard in quantitative NMR, GC-MS, and LC-MS workflows, providing consistent isotopic labeling for accurate spectral and mass analyses.
- Deuterium-labeled (d8) isotopologue for isotope-dilution and tracing experiments.
- High stated purity of 99.33% for reliable analytical performance.
- Molecular formula C10D8N2 and molecular weight 164.24 g·mol⁻¹.
- Soluble in DMSO (~100 mg/mL) with ultrasonic assistance for solution preparation.
- Stable when stored as a powder at -20°C for long-term storage or 4°C for shorter term.
- Suitable as an internal standard for quantitative NMR, GC-MS, and LC-MS analyses.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.