Learn More
Medchemexpress LLC 2 2-Bipyridine-d8 | MFCD00190432 | 99.3% | 164.24 g/mol | C10D8N2 | 25 MG
SDP
Supplier:  Medchemexpress LLC HYW44040925MG
Encompass_Preferred
2,2'-Bipyridine-d8 is the deuterium-labeled isotopologue of 2,2'-bipyridine supplied as a white to off-white solid. It is used as an isotopically labeled reagent or internal standard in organic synthesis, mechanistic studies, and mass spectrometry.
  • High isotopic enrichment suitable for analytical standards.
  • High purity (99.3%) for reliable research results.
  • Solid form enables accurate weighing and convenient storage.
  • Suitable as a ligand in coordination chemistry and mechanistic investigations.
  • Available in small, research-scale pack sizes for lab use.
Catalog No. 50-002-81169