Learn More
Medchemexpress LLC 2-[(2-chloro-4-fluorophenyl)methyl]-3-(4-fluorophenyl)-7-(trifluoromethyl)indazole | 875787-07-8 | MFCD16495812 | 100.0% | 422.78 g·mol⁻¹ | C21H12ClF5N2 | 200 MG
SDP
Supplier:  Medchemexpress LLC HY10629200MG
Encompass_Preferred
LXR-623 is a brain-penetrant liver X receptor (LXR) modulator that functions as a partial agonist at LXRα and a full agonist at LXRβ. It displays submicromolar potency and is provided as a high-purity research solid for studies of LXR signaling, CNS pharmacology, and cholesterol-related mechanisms.
  • Brain-penetrant compound suitable for CNS LXR studies.
  • Partial agonist at LXRα and full agonist at LXRβ.
  • Potent activity with IC50s of 24 nM (LXRβ) and 179 nM (LXRα).
  • Supplied as a high-purity analytical grade solid (>99%).
  • Useful for pharmacology, mechanism-of-action, and biomarker research.
Catalog No. 50-002-42254