Learn More
Medchemexpress LLC 2,2-dibenzyl-1,3-propanediol | 31952-16-6 | 99.4% | 256.35 | C17H20O2 | 5 G

Supplier: Medchemexpress LLC HYW0896975G
2,2-Dibenzyl-1,3-propanediol is a drug intermediate used in the synthesis of various active compounds. It is supplied for research use only and is not intended for human therapeutic use.
- Drug intermediate for synthesis applications.
- High purity (99.38%).
- Molecular formula C17H20O2.
- Molecular weight 256.35.
- Pack size 5 g for laboratory use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.