Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2,2-dibenzyl-1,3-propanediol | 31952-16-6 | MFCD01112173 | 99.4% | 256.35 g/mol | C17H20O2 | 250 MG
SDP

Supplier:  Medchemexpress LLC 250MGHYW089697

Encompass_Preferred

2,2-Dibenzyl-1,3-propanediol is a chemical intermediate used in medicinal chemistry for the synthesis of various active compounds. It is supplied for research use and typically offered at high purity for synthetic applications.

  • High purity (99.4%).
  • Molecular weight 256.35 g/mol.
  • Chemical formula C17H20O2.
  • Suitable as a drug intermediate for organic synthesis.
  • Available in multiple package sizes, including 250 MG.
  • Intended for research use only.

Catalog No. 50-002-08930


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.