Learn More
Medchemexpress LLC 2 2-Dimethoxy-2-phen 1kg
SDP
Supplier:  Medchemexpress LLC HYW0980211KG
Encompass_Preferred
2 2-Dimethoxy-2-phenylacetophenone (Photoinitiator-651) is a photoinitiator that by absorbing UV light generates free radicals to initiate cross-linking reactions thereby curing coating materials and forming adhesive coatings 2 2-Dimethoxy-2-phenylacetophenone can be used in research on the preparation of conductive adhesives 3D printing materials medical coatings and biocompatible glues for biomedical devices[1]
Catalog No. 50-002-05576