Learn More
Medchemexpress LLC 2,2'-Dipyridyl disulfide | 2127-03-9 | 99.98% | 220.31 | 50 G
SDP
Supplier:  Medchemexpress LLC 50GHYY1666
Encompass_Preferred
2,2'-Dipyridyl disulfide is a chemical reagent primarily utilized for the determination of sulfhydryl groups. It also serves as a common reagent in peptide chemistry, often employed in oxidation-reduction condensations to form peptide bonds or in coupling reactions to create disulfide-linked heterodimers.
  • Useful for the determination of sulfhydryl groups.
  • Facilitates oxidation-reduction condensations in peptide bond formation.
  • Enables coupling reactions to form disulfide-linked heterodimers.
  • Accelerates intramolecular S-S bond formation in cysteine-containing peptides.
  • Hazardous; harmful if swallowed and causes skin and serious eye irritation.
  • May cause respiratory irritation.
  • For research use only, requiring handling by qualified personnel in appropriate facilities.
Catalog No. 50-003-65927