Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2,2'-(Piperazine-1,4-diyl)diethanamine | 6531-38-0 | 172.27 | 50 MG
SDP

Supplier:  Medchemexpress LLC HYW00444050MG

Encompass_Preferred

2,2'-(Piperazine-1,4-diyl)diethanamine is a piperazine with two N-ethanamine groups. This molecule can be used to build drug molecules or design novel ionizable aminolipid head groups for lipid nanoparticles. This product is for research use only.

  • Solid appearance
  • Color: white to light yellow
  • Soluble in H2O at 10 mg/mL (58.05 mM; ultrasonic and warming and heat to 60°C)
  • Chemical formula: C8H20N4

Catalog No. 50-004-48884


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.