Learn More
Business Communication Solutions 2.25"W x 1.25"L, White Matte Polypropylene Direct Thermal Label, Permanent Adhesive, IPA Resistant Varnish, 610 Labels/Roll, 12 Rolls/Case, 0.75" Core, 2.8715" OD.
Supplier:  Business Communication Solutions B14380
Encompass_Preferred
2.25"W x 1.25"L, White Matte Polypropylene Direct Thermal Label. Offers the highest level of cleanliness available for today's critical environments. Gamma irradiated (dose level of 25-50kGy) backed with a permanent adhesive and an IPA resistant varnish, which helps protect the printed image when wiped down with IPA. Offers cleanroom packaging and is ideal for keeping products safe from excess environmental contamination prior to use. Cleaned with an in-line sub-micron, MicroClean 10, web cleaning system, which breaks down the static layer, allowing both sides of label stock to be cleaned ultrasonically. Packaged in a fully-ionized cleanroom in MDPE certified cleanroom bags, which are nitrogen purged, backfilled, and then vacuum sealed in double bags. This ensures that the labels are sealed in a sub-micron environment to maintain the cleanliness level and freshness of the labels. Compatible with Zebra Models ZQ620 and QLn320 Thermal Barcode Printer. Two year expiration.
Catalog No. NC2200728