Learn More
Medchemexpress LLC 2-[(2E)-2-(1-azabicyclo[2.2.2]oct-3-ylidene)-2-fluoroethoxy]-9H-carbazole HCl | 182959-33-7 | 99.4% | 372.86 g/mol | C21H22ClFN2O | 1 ML
SDP
Supplier:  Medchemexpress LLC HY100313A10MM 1ML
Encompass_Preferred
YM-53601 is a squalene synthase inhibitor used in biochemical and cellular research to probe cholesterol biosynthesis and lipid metabolism. It is provided as a dry solid or as a ready-to-use 10 mM solution in DMSO (1 mL), and is characterized by molecular formula C21H22ClFN2O, molecular weight 372.86 g/mol, and CAS 182959-33-7.
  • Squalene synthase inhibitor useful for cholesterol pathway studies.
  • Available as solid or 10 mM solution in DMSO for immediate use.
  • High purity suitable for biochemical and cell-based assays.
  • Includes structural identifiers for compound confirmation.
  • Storage recommendations provided to preserve stability.
Catalog No. 50-002-14202