Learn More
Pacer Global LLC Marathon LS 2.2mL V Bottom Deep Well Storage Plates, 96 Square Well, Non Sterile, 50 per Case
SDP
Supplier:  Pacer Global LLC MLSDSWP96V22/DEL
Encompass_Preferred
96 Well Deep Well Plate, working volume of 2.2 mL. Polypropylene. Square well with ‘V’ bottom. Minimal remaining volume and high uniformity from well to well. Raised well rims, smooth surface for reliable closing, even with heat sealing. Easy and reliable stacking. Good centrifugation stability up to 6,000xg for faster protocols and improved sample quality. Manufactured under DNase/ RNase free environment without slip agents, plasticizers or biocides – Materials which could have a negative effect on bioassays. Pure, virgin polypropylene guarantees good extractible performance, high resistance to chemicals, good mechanical stress and working with temperature extremes. 50/case
Catalog No. NC1862169