Learn More
Advanced Chemblocks Inc 2-3-3-DIFLUOROCYCLOBUTY 250MG
Supplier:  Advanced Chemblocks Inc P50164250MG
Encompass
2-(3,3-difluorocyclobutyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane (250MG)CAS: 2243345-24-4MDL: MFCD31705659Purity: 97%
Catalog No. NC3065424