Learn More
Cayman Chemical 2 3 3TrImethylIndolenIn5s
SDP
Supplier:  Cayman Chemical 17647100 MG
Encompass_Preferred
A water soluble, fluorescent near-infrared dye that can be conjugated to peptides, proteins, nucleic acids, RNA, DNA, carbohydrates, polymers, and small molecules via a sulfonyl substituent
Catalog No. 50-296-6573