Learn More
Medchemexpress LLC 2,3,4,5-Tetrahydro-1H-1,5-methano-3-benzazepine | 69718-72-5 | 99.78% | 159.23 | 10 G

Supplier: Medchemexpress LLC HY2000710G
2,3,4,5-Tetrahydro-1H-1,5-methano-3-benzazepine is a drug intermediate used for the synthesis of various active compounds. It is for research use only.
- Appearance: solid-liquid mixture
- Color: light yellow to brown
- Shipping: room temperature in continental US; may vary elsewhere
- Storage: 4°C, protect from light. In solvent: -80°C for 6 months; -20°C for 1 month (protect from light)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.