Learn More
Medchemexpress LLC 2,3,4,5-Tetrahydro-3-(trifluoroacetyl)-1,5-methano-1H-3-benzazepine-7,8-diamine | 230615-69-7 | 99.9% | 285.27 | 25 G

Supplier: Medchemexpress LLC 25GHY20010
This compound, 2,3,4,5-Tetrahydro-3-(trifluoroacetyl)-1,5-methano-1H-3-benzazepine-7,8-diamine, serves as a drug intermediate for the synthesis of various active compounds.
- High purity: 99.92%
- Appearance: solid
- Color: off-white to pink
- Targeted as a drug intermediate
- Recommended storage at 4°C, sealed, away from moisture and light
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.