Learn More
Medchemexpress LLC 2,3,4,5-Tetrahydro-3-(trifluoroacetyl)-1,5-methano-1H-3-benzazepine-7,8-diamine | 230615-69-7 | 99.92% | 285.27 | 500 MG

Supplier: Medchemexpress LLC HY20010500MG
This product is 2,3,4,5-Tetrahydro-3-(trifluoroacetyl)-1,5-methano-1H-3-benzazepine-7,8-diamine, an off-white to pink solid used as a drug intermediate for the synthesis of various active compounds. It is intended for research purposes only, not for sale to patients.
- Molecular weight: 285.27
- Molecular formula: C13H14F3N3O
- Purity: 99.92%
- Shipping: Room temperature in continental US; may vary elsewhere
- Storage: 4°C, sealed, away from moisture and light
- In solvent storage: -80°C for 6 months; -20°C for 1 month (sealed, away from moisture and light)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.