Learn More
Medchemexpress LLC 2-(3,4-dihydroxyphenyl)-2-oxoethyl ester thiocyanic acid | 101714-41-4 | MFCD22415334 | 99.6% | 209.22 g/mol | C9H7NO3S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1101885MG
Encompass_Preferred
BiP inducer X (BIX) is a small-molecule research reagent that selectively induces the endoplasmic reticulum chaperone BiP/GRP78. It is used to modulate ER stress responses and investigate protein folding, secretion, and apoptosis in cellular models.
  • Selective induction of BiP/GRP78 in vitro.
  • Enables study of ER stress and unfolded protein response pathways.
  • High purity for reliable experimental results.
  • Stable powder with defined storage conditions for long-term use.
  • Available in small research pack sizes for dose-ranging studies.
Catalog No. 50-002-63735