Learn More
Medchemexpress LLC 2-(3,4-dimethoxyphenyl)-5-fluorobenzothiazole | 872726-44-8 | MFCD12024619 | 99.8% | 289.32 g/mol | C15H12FNO2S | 50 MG
SDP
Supplier:  Medchemexpress LLC HY11847450MG
Encompass_Preferred
GW 610 is an aryl hydrocarbon receptor (AhR) ligand investigated as an antitumor research compound. It has demonstrated selective cytotoxicity in lung, colon, and breast cancer cell lines and induces CYP1A1 expression in MCF-7 cells.
  • Potent AhR ligand with reported Ki = 6.8 nM.
  • Demonstrated selective anticancer activity in lung, colon, and breast cancer cell lines.
  • Induces CYP1A1 expression in MCF-7 cells.
  • High purity, reported as 99.78% (certificate of analysis).
  • Molecular formula C15H12FNO2S; molecular weight 289.32 g/mol.
  • Intended for in vitro research use only; not for human or clinical use.
Catalog No. 50-002-28342