Learn More
Medchemexpress LLC HY-W016993 100mg Medchemexpress, 2,3,4-Trihydroxybenzoic acid CAS:610-02-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW016993100MG
Encompass_Preferred
Medchemexpress, HY-W016993 100mg 2,3,4-Trihydroxybenzoic acid CAS:610-02-6 2,3,4-Trihydroxybenzoic acid is an internal standard in separation of phenolic acids by HPLC. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4623