Learn More
Medchemexpress LLC 2,3,4-Trihydroxybenzophenone | 1143-72-2 | 99.9% | 5 G
SDP
Supplier:  Medchemexpress LLC HYD02725G
Encompass_Preferred
2,3,4-Trihydroxybenzophenone is a hydroxylated benzophenone UV filter that exhibits estrogenic activity. It is also a quorum sensing inhibitor and an EC 1.14.18.1 (tyrosinase) inhibitor. This compound is known to be a human urinary metabolite, a rat metabolite, and a drug metabolite.
  • Hydroxylated benzophenone UV filter
  • Exhibits estrogenic activity
  • Quorum sensing inhibitor
  • EC 1.14.18.1 (tyrosinase) inhibitor
  • Human urinary metabolite
  • Rat metabolite
  • Drug metabolite
Catalog No. 50-004-10286