Learn More
Medchemexpress LLC 2,3,4-Trihydroxybenzophenone | 1143-72-2 | MFCD00009996 | 99.9% | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYD0272
Encompass_Preferred
2,3,4-Trihydroxybenzophenone is a hydroxylated benzophenone UV filter known for its estrogenic and anticancer activity. It also acts as a quorum sensing inhibitor and an EC 1.14.18.1 (tyrosinase) inhibitor. This compound is identified as a human urinary, rat, and drug metabolite, and is relevant in metabolic and endocrine disease research.
  • Functions as a UV filter.
  • Exhibits estrogenic activity.
  • Acts as a quorum sensing inhibitor.
  • Inhibits EC 1.14.18.1 (tyrosinase).
  • Identified as a metabolite.
  • Involved in vitamin D related/nuclear receptor and estrogen receptor/ERR pathways.
Catalog No. 50-004-13934