Learn More
Medchemexpress LLC 2',3',4'-Trimethoxyacetophenone | 13909-73-4 | 97.0% | C11H14O4 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY7831510MM/1ML
Encompass_Preferred
2',3',4'-Trimethoxyacetophenone is an organic compound with a benzene ring and an acetyl group.
  • Can be used as a raw material for synthesizing dyes.
  • Can be used as a raw material for synthesizing fragrances.
  • Can be used as a raw material for synthesizing certain pesticides.
Catalog No. 50-004-33201