Learn More
Medchemexpress LLC 2,3,5,6-tetrachloro-1,4-benzoquinone | 118-75-2 | MFCD00001594 | 98.0% | 245.9 g/mol | C6Cl4O2 | 250g
SDP
Supplier:  Medchemexpress LLC HYY0278250G
Encompass_Preferred
Chloranil (Tetrachloro-p-benzoquinone) an orally active metabolite of pentachlorophenol and hexachlorobenzene is a widely used fungicide Chloranil can induce ROS production Chloranil induces neutrophil extracellular traps through the ROS-JNK-NOX2 pathway Chloranil induces ferroptosis and neuroinflammation Chloranil induces apoptosis of mouse embryonic stem cells[1][2][3][4][5]
Catalog No. 50-002-69980