Learn More
Medchemexpress LLC 2-(3,5-dimethylpyrazol-1-yl)-5-methylphenol | 1391758-52-3 | 100.0% | 202.25 g/mol | C12H14N2O | 25 MG
SDP
Supplier:  Medchemexpress LLC HY10801225MG
Encompass_Preferred
ME1111 is a small-molecule antifungal research compound that inhibits succinate dehydrogenase in Trichophyton species and is used in onychomycosis studies due to its ability to penetrate human nails. The compound is a phenolic pyrazole derivative with formula C12H14N2O and CAS 1391758-52-3.
  • Inhibits fungal succinate dehydrogenase.
  • Active against dermatophytes.
  • Demonstrates good human nail penetration for onychomycosis research.
  • Small-molecule phenolic pyrazole with formula C12H14N2O.
  • High purity suitable for in vitro and in vivo research applications.
Catalog No. 50-002-42174