Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2,3,5-tri-O-benzyl-D-ribono-1,4-lactone | 55094-52-5 | MFCD08703966 | 100.0% | 418.49 g/mol | C26H26O5 | 1 ML
SDP

Supplier:  Medchemexpress LLC HYW01860410MM/1ML

Encompass_Preferred

2,3,5-Tri-O-benzyl-D-ribono-1,4-lactone is a benzyl-protected D-ribono-1,4-lactone provided as a ready-to-use 10 mM solution in DMSO. It is commonly used as a protected sugar intermediate in nucleoside and carbohydrate synthesis and related organic chemistry applications. The material is supplied at high purity for research use.

  • Ready-to-use 10 mM solution in DMSO.
  • High purity (99.95%).
  • Molecular formula C26H26O5; molecular weight 418.49 g/mol.
  • White to off-white solid appearance; supplied as a solution for convenience.
  • Storage stability in solvent: -80°C for 6 months, -20°C for 1 month.
  • Suitable as a protected sugar intermediate for synthetic chemistry.

Catalog No. 50-003-00844


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.