Learn More
Medchemexpress LLC 2,3,5-tri-O-benzyl-D-ribono-1,4-lactone | 55094-52-5 | MFCD08703966 | 100.0% | 418.49 g/mol | C26H26O5 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW01860410G
Encompass_Preferred
2,3,5-Tri-O-benzyl-D-ribono-1,4-lactone is a benzyl-protected ribonolactone used as a synthetic intermediate in carbohydrate and nucleoside chemistry. It is supplied as a stable powder suitable for laboratory synthesis and research applications involving protected ribose derivatives and nucleoside analogs.
  • High purity suitable for synthetic applications.
  • Benzyl-protected sugar scaffold for selective transformations.
  • Stable powder with defined storage conditions for longevity.
  • Useful for nucleoside analog synthesis and carbohydrate chemistry.
  • Available in multiple pack sizes for scale flexibility.
Catalog No. 50-003-01063