Learn More
Medchemexpress LLC 2,3,5-tri-O-benzyl-D-ribono-1,4-lactone | 55094-52-5 | MFCD08703966 | 100.0% | 418.49 g/mol | C26H26O5 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0186045G
Encompass_Preferred
2,3,5-Tri-O-benzyl-D-ribono-1,4-lactone is a benzyl-protected D-ribono-1,4-lactone used as a protected sugar intermediate and building block in the synthesis of nucleoside analogs and other organic compounds. It has formula C26H26O5, a molecular weight of 418.49 g/mol, and is supplied at high purity for laboratory research.
Catalog No. 50-003-02008