Learn More
Medchemexpress LLC 2,3,5-tri-O-benzyl-D-ribose | 54623-25-5 | MFCD03425640 | >95.0% | 420.5 g/mol | C26H28O5 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW03720010G
Encompass_Preferred
2,3,5-Tri-O-benzyl-D-ribose is a benzyl-protected D-ribose derivative used as a synthetic building block in organic and medicinal chemistry. It functions as a protected sugar intermediate for multi-step syntheses, enabling selective transformations of ribose derivatives. Molecular formula C26H28O5, molecular weight 420.5 g/mol.
  • Protects hydroxyl groups during multi-step synthesis
  • Suitable for carbohydrate and nucleoside synthesis
  • Stable under common organic reaction conditions
  • Offered in multiple laboratory pack sizes for scalability
  • Useful intermediate for preparing ribose-containing derivatives
Catalog No. 50-003-01199