Learn More
Medchemexpress LLC 2,3,5-tri-O-benzyl-D-ribose | 54623-25-5 | MFCD03425640 | 98.6% | 420.5 | C26H28O5 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW037200
Encompass_Preferred
2,3,5-Tri-O-benzyl-D-ribose is a benzyl-protected derivative of D-ribose supplied as a powder for research use. It is used as an intermediate in carbohydrate and nucleoside synthesis and in organic synthesis workflows that require protected sugars. The material is characterized for purity and accompanied by storage recommendations to preserve stability.
  • High purity: 98.6%.
  • Confirmed molecular formula C26H28O5 and molecular weight 420.5.
  • Powder form for easy weighing and dissolution.
  • Suitable as a synthetic intermediate in carbohydrate and nucleoside chemistry.
  • Store at -20°C under inert atmosphere to maintain stability.
Catalog No. 50-003-03026