Learn More
Medchemexpress LLC 2,3-Bis(3-indolylmethyl)indole | 138250-72-3 | 375.47 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYN1011710MM/1ML
Encompass_Preferred
2,3-Bis(3-indolylmethyl)indole significantly suppresses RANKL-induced osteoclast formation, actin ring formation, and bone resorption in a concentration-dependent manner.
  • Significantly suppresses RANKL-induced osteoclast formation
  • Inhibits actin ring formation
  • Reduces bone resorption
  • High purity (99.53%)
  • Light yellow to light brown solid appearance
  • Soluble in DMSO
Catalog No. 50-004-17158