Learn More
Medchemexpress LLC 2-(3-chloro-4-fluorophenyl)-N-(1-methylethyl)-6-(3-morpholinylpropoxy)pyridopyrimidine hydrochloride | 1642187-96-9 | 99.7% | 554.44 g·mol⁻¹ | C25H30Cl2FN5O4 | 25 MG

Supplier: Medchemexpress LLC HY11782025MG
TASP0390325 is a high-affinity, orally active arginine vasopressin V1B receptor antagonist used in preclinical pharmacology studies. It demonstrates antidepressant and anxiolytic activity in rodent models and antagonizes CRF/dDAVP-induced ACTH release. The compound is supplied as a white to off-white solid with high reported purity and is commonly handled as a research chemical.
- High-affinity V1B receptor antagonist.
- Orally active with demonstrated antidepressant and anxiolytic effects in rodent models.
- High purity (>99%) white to off-white solid.
- Soluble in DMSO (≈50 mg/mL); practically insoluble in water.
- Recommended storage: sealed, away from moisture; -20°C for solid; in solution keep at -80°C (6 months) or -20°C (1 month).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.