Learn More
Medchemexpress LLC 2,3-Diaminopropanoic acid hydrochloride | 54897-59-5 | ≥98.0% | 140.57 | 10 G

Supplier: Medchemexpress LLC HYW01367410G
2,3-Diaminopropanoic acid hydrochloride is an endogenous metabolite.
- Endogenous metabolite.
- Purity of 98.0%.
- Available in multiple solid package sizes.
- High solubility in water (100 mg/mL).
- Recommended storage for solid form is 4°C, sealed and away from moisture.
- For solutions, storage at -80°C for 6 months or -20°C for 1 month is recommended.
- Intended for research use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.