Learn More
Medchemexpress LLC 2,3-Diaminopropanoic acid hydrochloride | 54897-59-5 | 98.0% | 140.57 | 25 G

Supplier: Medchemexpress LLC 25GHYW013674
2,3-Diaminopropanoic acid hydrochloride is an endogenous metabolite. It is for research use only and not sold to patients.
- Endogenous metabolite
- For research use only
- Available in various quantities (e.g., 500 mg, 1 g, 5 g, 10 g, 25 g)
- Soluble in H2O (100 mg/mL)
- Purity: 98.0%
- Appearance: Solid
- Color: White to off-white
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.