Learn More
Medchemexpress LLC 2,3-Dichloropyridine | 2402-77-9 | 99.96% | 147.99 | 10 MM 1 ML

Supplier: Medchemexpress LLC HYW01259110MM/1ML
2,3-Dichloropyridine is a biochemical reagent that can be used as a biological material or organic compound for life science related research.
- High purity: 99.96%
- Versatile usage: suitable for various life science applications
- Multiple forms available: powder and in solvent
- Well-documented: data sheets, SDS, and handling instructions available
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.