Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2',3'-dideoxy-5-iodocytidine | 114748-57-1 | 99.3% | 337.11 g/mol | C9H12IN3O3 | 25 MG
SDP

Supplier:  Medchemexpress LLC 25MGHYW048478

Encompass_Preferred

2',3'-dideoxy-5-iodocytidine is an iodinated nucleoside analogue used as a chain-terminating reagent in DNA sequencing and as a research antibiotic with reported activity against Mycobacterium. It is supplied as a solid for laboratory research applications.

  • Used as a chain-terminating reagent for DNA sequencing.
  • Reported antibiotic activity against Mycobacterium.
  • Molecular formula C9H12IN3O3.
  • Molecular weight 337.11 g/mol.
  • CAS number 114748-57-1.
  • Available as powder in 25 MG to 500 MG pack sizes.

Catalog No. 50-002-09623


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.