Learn More
Medchemexpress LLC 2',3'-Dideoxyadenosine 5'-triphosphate (trisodium) | 72029-21-1 | 99.9% | 54 MG
SDP
Supplier:  Medchemexpress LLC HY128036B54MG
Encompass_Preferred
ddATP (2',3'-Dideoxyadenosine 5'-triphosphate) trisodium is an active metabolite of 2',3'-dideoxyinosine. It functions as a chain-elongating inhibitor of DNA polymerase and is useful for various applications.
  • Inhibitor of DNA polymerase
  • Used for Sanger method for DNA sequencing
  • Aids in research of virus infection
  • Inhibits adenovirus DNA synthesis
  • Reduces γH2A.X foci in DSB oocytes
  • Inhibits HIV replication by inhibiting reverse transcriptase
Catalog No. 50-003-42341