Learn More
Cayman Chemical 2 3-DIdeoxygunosIn-5-O-t 2.5mg
SDP
Supplier:  Cayman Chemical 383752.5 MG
Encompass_Preferred
A reverse transcriptase inhibitor (Kis = 13 and 6.7 nM for the visna virus and HIV-1 enzymes, respectively); has been used in the synthesis of oligonucleotides containing a terminal ddG that are resistant to cleavage by SARS-CoV-2 exonuclease; has been used to terminate chain extension produced by the Taq polymerase in PCR assays
Catalog No. 50-291-6697