Learn More
Medchemexpress LLC 2,3-Dimercapto-1-propanol | 59-52-9 | 98.0% | 500 MG
SDP
Supplier:  Medchemexpress LLC HYB1285500MG
Encompass_Preferred
Dimercaprol (2,3-Dimercapto-1-propanol) is an anti-heavy metal-poisoning agent that also exhibits anti-HIV activity. It can be utilized for research into poisoning caused by arsenic, mercury, gold, lead, antimony, and other toxic metals.
  • Anti-heavy metal-poisoning agent
  • Exhibits anti-HIV activity
  • Useful for studying poisoning from arsenic, mercury, gold, lead, and antimony
  • Significantly protected PC-12 cells from Acrolein-mediated cell death in a dose-dependent manner in vitro.
  • Decreased mercury contents in the liver and kidney in Wistar rats in vivo.
Catalog No. 50-004-12842