Learn More
Cayman Chemical 2 3-DImercaptopropnsulfonat 1g
SDP
Supplier:  Cayman Chemical 395211 G
Encompass_Preferred
A mercury chelator; reduces renal mercury levels and increases urinary mercury levels in uninephrectomized- and normal rats administered mercuric chloride; inhibits methylmercury chloride-induced increases in embryotoxicity and fetal- and maternal death and decreases in fetal bone ossification in a mouse model of heavy metal-induced developmental toxicity when administered to pregnant mice at 180 or 360 mg/kg per day for four days
Catalog No. 50-291-7581