Learn More
Medchemexpress LLC 2,3-diphenylquinoxaline-6-carboxylic acid | 32387-96-5 | MFCD00555042 | 100.0% | 326.35 | C21H14N2O2 | 500 MG
SDP
Supplier:  Medchemexpress LLC HYW028714500MG
Encompass_Preferred
2,3-Diphenylquinoxaline-6-carboxylic acid is a quinoxaline-derived carboxylic acid provided for research use as a chain-end modifier. It is primarily employed to end-cap AB2 monomers and related synthetic intermediates, and is supplied with analytical documentation supporting identity and purity.
  • Used as a chain-end modifier in end-capping of AB2 monomers
  • Applicable to synthesis of polymer building blocks and intermediates
  • Molecular formula C21H14N2O2 and molecular weight 326.35
  • High purity (≈99.95% by LCMS) suitable for research applications
  • Available in small-scale laboratory packaging (500 mg)
Catalog No. 50-002-99337