Learn More
Medchemexpress LLC 2-[3-[(E)-hydroxyiminomethyl]-4-(2-methylpropoxy)phenyl]-4-methyl-1,3-thiazole-5-carboxylic acid | 1350352-70-3 | MFCD31807379 | 98.2% | C16H18N2O4S | 10MG
SDP
Supplier:  Medchemexpress LLC HY13126910MG
Encompass_Preferred
Febuxostat impurity 7 is an analytical reference standard representing a specific impurity of febuxostat (CAS 1350352-70-3). It is supplied as a characterized solid for research and analytical applications including impurity profiling, method development, and quality control. The compound has molecular formula C16H18N2O4S and molecular weight 334.39.
  • High purity (98.2%).
  • Characterized identity for reliable reference standard use.
  • Suitable for analytical methods such as HPLC and LC-MS.
  • Intended for impurity profiling, method development, and quality control.
  • Provided as a stable solid for laboratory handling and storage.
Catalog No. 50-002-61982