Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2',3'-O-isopropylidene-6-mercaptopurine riboside | 5856-48-4 | MFCD00051198 | 96.6% | 324.36 | C13H16N4O4S | 50 MG
SDP

Supplier:  Medchemexpress LLC HYW55599150MG

Encompass_Preferred

2',3'-O-isopropylidene-6-mercaptopurine riboside is an isopropylidene-protected nucleoside derivative used as a research reagent with notable metal complexing activity. It is supplied as a solid for laboratory use and should be handled and stored under appropriate conditions to preserve stability.

  • CAS number 5856-48-4.
  • Molecular formula C13H16N4O4S.
  • Molecular weight 324.36.
  • Purity approximately 96.6%.
  • Packaged quantity 50 MG.
  • Appearance solid, white to off-white.
  • Storage: powder at -20°C up to 3 years; in solvent -80°C for 6 months or -20°C for 1 month.
  • Application for studying metal complexing mechanisms in research.

Catalog No. 50-002-84129


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.