Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2,3-Pentanedione | 600-14-6 | 99.7% | 100.12 | 500 G
SDP

Supplier:  Medchemexpress LLC HYW012998500G

Encompass_Preferred

2,3-Pentanedione is a common component of synthetic flavorings. It is used to provide a butter, strawberry, caramel, fruit, rum, or cheese flavor in various products, including beverages, ice cream, candy, baked goods, gelatins, and puddings. This substance also occurs naturally as a fermentation product in beer, wine, and yogurt, and is released during the roasting of coffee beans. The product is intended for research use only and is not sold to patients.

  • Flavoring agent: Imparts various desirable flavors (butter, strawberry, caramel, fruit, rum, cheese) to food and beverage products.
  • Naturally occurring: Found as a fermentation product in beer, wine, and yogurt, and released during coffee bean roasting.
  • High purity: 99.74% purity ensures reliability for research applications.
  • Versatile applications: Applicable in a wide range of food and beverage categories.

Catalog No. 50-004-51392


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.