Learn More
Medchemexpress LLC 2(3h)-Furanone, dihydro-5-[(3-hydroxyphenyl)methyl]- | 21618-91-7 | 99.9% | 192.21 | 10 MG

Supplier: Medchemexpress LLC 10MGHYW165608
5-(3'-Hydroxyphenyl)-γ-valerolactone is an endogenous flavan-3-ols metabolite found in human faecal microbiota.
- White to off-white solid appearance
- Purity of 99.9%
- Store as powder at -20°C for up to 3 years
- Store in solvent at -80°C for up to 6 months
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.