Learn More
Cayman Chemical 2 3IsopropylIdenDrIbonla 500mg
SDP
Supplier:  Cayman Chemical 30479500 MG
Encompass_Preferred
A building block; has been used in the stereocontrolled synthesis of 6-epi-trehazolin and 6-epi-trehalamine
Catalog No. 50-299-2000