Learn More
Medchemexpress LLC 2-[4-(1-aminocyclobutyl)phenyl]-3-phenylimidazo[1,2-b]pyridazine-6-carboxamide | 1402608-02-9 | MFCD30481340 | 99.5% | 383.4 g/mol | C23H21N5O | 50 MG
SDP
Supplier:  Medchemexpress LLC HY10001850MG
Encompass_Preferred
BAY1125976 is a selective, allosteric small-molecule inhibitor of AKT isoforms 1 and 2 used as a research probe to study PI3K/AKT/mTOR signaling and AKT-dependent cancer biology. It is orally bioavailable and demonstrates potent biochemical activity in the low nanomolar range.
  • Selective allosteric inhibition of AKT1 and AKT2.
  • Potent biochemical activity at low nanomolar concentrations.
  • Suitable for biochemical and cellular assays.
  • Provided with quality documentation such as COA and LC-MS.
  • Solid powder with recommended storage to preserve stability.
  • High purity suitable for research applications.
Catalog No. 50-002-15347