Learn More
Medchemexpress LLC 2,4(1H,3H)-Pyrimidinedione, 5-methyl- | 65-71-4 | 100.0% | 126.11 | 50 G
SDP
Supplier:  Medchemexpress LLC HYW01045050G
Encompass_Preferred
Thymine is one of the four bases of DNA, acting as a substrate for rat liver dihydropyrimidine dehydrogenase (DPD). This product is suitable for research in endogenous metabolite, metabolic disease, and cancer. It appears as a white to off-white solid.
  • Consistent with structure based on 1 H NMR Spectrum
  • Consistent with structure based on LCMS
  • Purity of 99.98% via LCMS
  • Product has been tested and complies with specifications
Catalog No. 50-004-53789