Learn More
Medchemexpress LLC 2,4(1H,3H)-Pyrimidinedione-6-d, 5-(methyl-d3)- | 200496-79-3 | 98% | 130.14 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYW010450S
Encompass_Preferred
Thymine-d4 is the deuterium labeled Thymine, one of the four nucleobases in the nucleic acid of DNA. It can be a target for actions of 5-fluorouracil (5-FU) in cancer treatment, with a Km of 2.3 μM. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules as tracers for quantitation during drug development. Deuteration, in particular, has potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-65792