Learn More
Medchemexpress LLC 2-(4-(2,3,4-trimethoxybenzyl)piperazin-1-yl)ethyl nicotinate | 2254741-41-6 | 99.1% | 415.48 g·mol⁻¹ | C22H29N3O5 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1395775MG
Encompass_Preferred
Ninerafaxstat is an orally active cardiac mitotrope used for research on cardiac energetics. It partially inhibits fatty acid oxidation by targeting 3-ketoacyl-CoA thiolase, shifting myocardial substrate utilization toward glucose oxidation and improving cardiac energy efficiency. Supplied as a high-purity research compound with documented molecular properties and formulation for laboratory use.
  • Partial fatty acid oxidation inhibition improves energy efficiency.
  • Oral bioactivity supports in vivo research models.
  • High reported purity suitable for experimental use.
  • Documented molecular weight and formula for analytical characterization.
  • Applicable to studies of cardiac metabolism and disease models.
Catalog No. 50-002-23142